siRNA Details
Gene Symbol: 5730457N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038163.1 | 4 | 2884 | |||
GenBank mRNA | AK031498.1 | - | 4 | 2886 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5730457N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038163.1 | 4 | 2884 | |||
GenBank mRNA | AK031498.1 | - | 4 | 2886 |