siRNA Details
| Gene Symbol: 5730457N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038163.1 | 1 | 13 | |||
| GenBank mRNA | AK031498.1 | - | 1 | 15 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB620958.1 | - | 1 | 15 | Chr. 6 - 52310648 - 52314832 [-] on Build GRCm38 | |

5730457N03Rik
Genomic Map