siRNA Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048798.1 | - | 1153 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hif1a| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048798.1 | - | 1153 | |||