siRNA Details
| Gene Symbol: 2210406O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043305.1 | - | 2 | 701 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB258116.2 | - | 658 | Chr. - - [-] on Build | ||

2210406O10Rik
Genomic Map