siRNA Details
| Gene Symbol: A730082K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040317.1 | 4 | 1636 | |||
| NR_040319.1 | 5 | 1614 | ||||
| GenBank mRNA | AK043303.1 | - | 2 | 1184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730082K24Rik| Gene Symbol: A730082K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040317.1 | 4 | 1636 | |||
| NR_040319.1 | 5 | 1614 | ||||
| GenBank mRNA | AK043303.1 | - | 2 | 1184 | ||