siRNA Details
| Gene Symbol: A730082K24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040317.1 | 4 | 1153 | |||
| NR_040319.1 | 5 | 1131 | ||||
| GenBank mRNA | AK043303.1 | - | 2 | 701 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV661602.1 | - | 1 | 114 | Chr. 7 - 114983192 - 114983960 [+] on Build GRCm38 | |

A730082K24Rik
Genomic Map