siRNA Details
| Gene Symbol: Eed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016228.1 | - | 1 | 706 | ||
| AK053947.1 | - | 700 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eed| Gene Symbol: Eed | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016228.1 | - | 1 | 706 | ||
| AK053947.1 | - | 700 | ||||