siRNA Details
| Gene Symbol: 2700022O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012274.1 | - | 1 | 843 | ||
| AV124825.1 | - | 1 | 222 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ565818.1 | - | 1 | 333 | Chr. 14 - 56667082 - 56667603 [-] on Build GRCm38 | |

2700022O18Rik
Genomic Map