siRNA Details
| Gene Symbol: 4930565D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040752.1 | 1 | 173 | |||
| GenBank mRNA | AK016226.1 | - | 1 | 174 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715834.1 | - | 1 | 174 | Chr. 3 - 84349575 - 84379921 [-] on Build GRCm38 | |

4930565D16Rik
Genomic Map