siRNA Details
| Gene Symbol: 4933431G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045163.1 | 2 | 261 | |||
| NR_045165.1 | 3 | 439 | ||||
| GenBank mRNA | AK016225.1 | - | 2 | 264 | ||
| AK017004.1 | - | 3 | 440 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715833.1 | - | 2 | 264 | Chr. 6 - 72120545 - 72122940 [-] on Build GRCm38 | |

4933431G14Rik
Genomic Map