siRNA Details
Gene Symbol: 4933431G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045163.1 | 2 | 374 | |||
NR_045165.1 | 3 | 552 | ||||
GenBank mRNA | AK016225.1 | - | 2 | 377 | ||
AK017004.1 | - | 3 | 553 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715833.1 | - | 2 | 377 | Chr. 6 - 72120545 - 72122940 [-] on Build GRCm38 |
