siRNA Details
| Gene Symbol: 4933431G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045163.1 | 3 | 898 | |||
| NR_045165.1 | 3 | 1302 | ||||
| GenBank mRNA | AK016225.1 | - | 3 | 901 | ||
| AK017004.1 | - | 3 | 1303 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933431G14Rik| Gene Symbol: 4933431G14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045163.1 | 3 | 898 | |||
| NR_045165.1 | 3 | 1302 | ||||
| GenBank mRNA | AK016225.1 | - | 3 | 901 | ||
| AK017004.1 | - | 3 | 1303 | |||