siRNA Details
| Gene Symbol: Gm16731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK164782.1 | - | 392 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730861.1 | - | 3 | 388 | Chr. 4 - 46535904 - 46539402 [+] on Build GRCm38 | |

Gm16731
Genomic Map