siRNA Details
| Gene Symbol: 1700026H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138516.1 | - | 1 | 284 | ||
| AK138667.1 | - | 1 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700026H06Rik| Gene Symbol: 1700026H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138516.1 | - | 1 | 284 | ||
| AK138667.1 | - | 1 | 284 | |||