siRNA Details
| Gene Symbol: 5430416B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017313.1 | - | 1 | 628 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430416B10Rik| Gene Symbol: 5430416B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017313.1 | - | 1 | 628 | ||