siRNA Details
| Gene Symbol: 9430095M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020525.1 | - | 1 | 345 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721949.1 | - | 1 | 342 | Chr. 4 - 55020913 - 55021286 [+] on Build GRCm38 | |

9430095M17Rik
Genomic Map