siRNA Details
| Gene Symbol: 9430093N23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020520.1 | - | 1 | 86 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721935.1 | - | 1 | 82 | Chr. 2 - 169234993 - 169237780 [+] on Build GRCm38 | |

9430093N23Rik
Genomic Map