siRNA Details
Gene Symbol: Olfr1077-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033507.1 | 2 | 524 | |||
GenBank mRNA | AF178753.1 | - | 1 | 106 | ||
DR065539.1 | - | 2 | 524 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1077-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033507.1 | 2 | 524 | |||
GenBank mRNA | AF178753.1 | - | 1 | 106 | ||
DR065539.1 | - | 2 | 524 |