siRNA Details
| Gene Symbol: Olfr1077-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF178753.1 | - | 1 | 19 | ||
| Gene Symbol: Olfr1065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF178750.1 | - | 1 | 19 | ||

Olfr1077-ps1
Genomic Map