siRNA Details
Gene Symbol: Gm31152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141156.1 | - | 132 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ062664.1 | - | 1 | 132 | Chr. 7 - 12866486 - 12868583 [-] on Build GRCm38 |
