siRNA Details
| Gene Symbol: Zfp664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035022.1 | - | 529 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB627255.1 | - | 4 | 529 | Chr. 5 - 124862764 - 124901557 [+] on Build GRCm38 | |

Zfp664
Genomic Map