siRNA Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036610.1 | - | 3162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd9| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036610.1 | - | 3162 | |||