siRNA Details
| Gene Symbol: E130114A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021388.1 | - | 1 | 479 | ||
| AK037750.1 | - | 1 | 2505 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130114A11Rik| Gene Symbol: E130114A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021388.1 | - | 1 | 479 | ||
| AK037750.1 | - | 1 | 2505 | |||