siRNA Details
| Gene Symbol: Fam109a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC095929.1 | - | 32 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW211627.1 | - | 1 | 29 | Chr. 5 - 121848208 - 121854298 [+] on Build GRCm38 | |

Fam109a
Genomic Map