siRNA Details
| Gene Symbol: 1700015G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005994.1 | - | 3 | 147 | ||
| AV042221.2 | - | 1 | 30 | |||
| AV042559.2 | - | 1 | 36 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700015G11Rik| Gene Symbol: 1700015G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005994.1 | - | 3 | 147 | ||
| AV042221.2 | - | 1 | 30 | |||
| AV042559.2 | - | 1 | 36 | |||