siRNA Details
| Gene Symbol: D930030I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086460.1 | - | 2 | 652 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB525975.2 | - | 2 | 426 | Chr. 16 - 38452207 - 38453493 [+] on Build GRCm38 | |

D930030I03Rik
Genomic Map