siRNA Details
| Gene Symbol: Gm26717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146643.1 | - | 111 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY739273.1 | - | 1 | 111 | Chr. 18 - 32153476 - 32159589 [+] on Build GRCm38 | |

Gm26717
Genomic Map