siRNA Details
| Gene Symbol: Utrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041435.1 | - | 1 | 762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Utrn| Gene Symbol: Utrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041435.1 | - | 1 | 762 | ||