siRNA Details
| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030934.1 | - | 1 | 2120 | ||
| AK037254.1 | - | 1 | 2126 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX515655.1 | - | 1 | 65 | Chr. 1 - 165707154 - 165707671 [-] on Build GRCm38 | |

Rcsd1
Genomic Map