siRNA Details
| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020768.1 | - | 2 | 1040 | ||
| AK030934.1 | - | 1 | 1125 | |||
| AK037254.1 | - | 1 | 1131 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcsd1| Gene Symbol: Rcsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020768.1 | - | 2 | 1040 | ||
| AK030934.1 | - | 1 | 1125 | |||
| AK037254.1 | - | 1 | 1131 | |||