siRNA Details
| Gene Symbol: BC035044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088258.1 | - | 3 | 540 | ||
| BC035044.1 | - | 3 | 696 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY746287.1 | - | 3 | 540 | Chr. 6 - 128849803 - 128890965 [-] on Build GRCm38 | |

BC035044
Genomic Map