siRNA Details
| Gene Symbol: BC035044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088258.1 | - | 3 | 979 | ||
| BC035044.1 | - | 3 | 1135 | |||
| Gene Symbol: 5830444B04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK167606.1 | - | 724 | |||

BC035044
Genomic Map