siRNA Details
| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076752.1 | - | 3 | 323 | ||
| AK161431.1 | - | 3 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700026D11Rik| Gene Symbol: 1700026D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076752.1 | - | 3 | 323 | ||
| AK161431.1 | - | 3 | 377 | |||