siRNA Details
| Gene Symbol: Pim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047301.1 | - | 914 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE849017.1 | - | 1 | 18 | Chr. 17 - 29463303 - 29464608 [-] on Build GRCm38 | |

Pim1
Genomic Map