siRNA Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011205.1 | - | 1 | 54 | ||
| AK143579.1 | - | 1 | 947 | |||
| AK149041.1 | - | 1 | 539 | |||
| AK169352.1 | - | 1 | 953 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbms1| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011205.1 | - | 1 | 54 | ||
| AK143579.1 | - | 1 | 947 | |||
| AK149041.1 | - | 1 | 539 | |||
| AK169352.1 | - | 1 | 953 | |||