siRNA Details
| Gene Symbol: 1110015O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045272.1 | 4 | 611 | |||
| GenBank mRNA | AK003731.2 | - | 1 | 127 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110015O18Rik| Gene Symbol: 1110015O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045272.1 | 4 | 611 | |||
| GenBank mRNA | AK003731.2 | - | 1 | 127 | ||