siRNA Details
| Gene Symbol: 1110015O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045272.1 | 4 | 836 | |||
| GenBank mRNA | AK003731.2 | - | 1 | 352 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703991.1 | - | 1 | 350 | Chr. 3 - 4813865 - 4814786 [+] on Build GRCm38 | |

1110015O18Rik
Genomic Map