siRNA Details
| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043828.1 | - | 3 | 1192 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB269030.2 | - | 3 | 632 | Chr. 12 - 33050786 - 33052419 [-] on Build GRCm38 | |

Cdhr3
Genomic Map