siRNA Details
| Gene Symbol: LOC102634401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110447.1 | 2 | 608 | |||
| GenBank mRNA | AK144266.1 | - | 2 | 609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102634401| Gene Symbol: LOC102634401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110447.1 | 2 | 608 | |||
| GenBank mRNA | AK144266.1 | - | 2 | 609 | ||