siRNA Details
| Gene Symbol: LOC102634401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110447.1 | 1 | 87 | |||
| GenBank mRNA | AK144266.1 | - | 1 | 88 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY733438.1 | - | 1 | 86 | Chr. 2 - 74752685 - 74753539 [+] on Build GRCm38 | |

LOC102634401
Genomic Map