siRNA Details
| Gene Symbol: 4930528J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015928.1 | - | 1 | 75 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715487.1 | - | 1 | 75 | Chr. 6 - 29915311 - 29915926 [+] on Build GRCm38 | |

4930528J11Rik
Genomic Map