siRNA Details
| Gene Symbol: Gm34284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132932.1 | - | 2, 3 | 271 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047210.1 | - | 2, 3 | 271 | Chr. 15 - 99068138 - 99074669 [-] on Build GRCm38 | |

Gm34284
Genomic Map