siRNA Details
| Gene Symbol: 9430024F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020436.1 | - | 1 | 850 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB367970.2 | - | 1 | 301 | Chr. 12 - 110590550 - 110591033 [-] on Build GRCm38 | |

9430024F10Rik
Genomic Map