siRNA Details
| Gene Symbol: A330049N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040646.1 | 5 | 1154 | |||
| GenBank mRNA | AK039487.1 | - | 5 | 1155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330049N07Rik| Gene Symbol: A330049N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040646.1 | 5 | 1154 | |||
| GenBank mRNA | AK039487.1 | - | 5 | 1155 | ||