siRNA Details
| Gene Symbol: A330049N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040646.1 | 4 | 378 | |||
| GenBank mRNA | AK039487.1 | - | 4 | 379 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB633452.1 | - | 4 | 379 | Chr. 10 - 72977663 - 73086705 [-] on Build GRCm38 | |

A330049N07Rik
Genomic Map