siRNA Details
Gene Symbol: A330049N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040646.1 | 4 | 479 | |||
GenBank mRNA | AK039487.1 | - | 4 | 480 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330049N07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040646.1 | 4 | 479 | |||
GenBank mRNA | AK039487.1 | - | 4 | 480 |