siRNA Details
| Gene Symbol: A330048O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045162.1 | 2 | 163 | |||
| GenBank mRNA | AK039480.1 | - | 2 | 153 | ||
| AK079799.1 | - | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330048O09Rik| Gene Symbol: A330048O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045162.1 | 2 | 163 | |||
| GenBank mRNA | AK039480.1 | - | 2 | 153 | ||
| AK079799.1 | - | 2 | 164 | |||