siRNA Details
| Gene Symbol: Gm20745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045745.1 | 3 | 1559 | |||
| GenBank mRNA | AK045659.1 | - | 3 | 1559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20745| Gene Symbol: Gm20745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045745.1 | 3 | 1559 | |||
| GenBank mRNA | AK045659.1 | - | 3 | 1559 | ||