siRNA Details
| Gene Symbol: Gm20745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045745.1 | 1 | 133 | |||
| GenBank mRNA | AK045659.1 | - | 1 | 134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB644171.1 | - | 1 | 131 | Chr. 9 - 68024699 - 68025712 [-] on Build GRCm38 | |

Gm20745
Genomic Map