siRNA Details
| Gene Symbol: Gm6329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040690.1 | 3 | 835 | |||
| GenBank mRNA | AK144087.1 | - | 3 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6329| Gene Symbol: Gm6329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040690.1 | 3 | 835 | |||
| GenBank mRNA | AK144087.1 | - | 3 | 837 | ||